missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trace Amine Receptor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24720SS
This item is not returnable.
View return policy
Description
Trace Amine Receptor 1 Polyclonal specifically detects Trace Amine Receptor 1 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Trace Amine Receptor 1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 86% homologous in mouse and 80% homologous in dog. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Q96RJ0 | |
| TAAR1 | |
| A portion of amino acids 225-250 of human TAAR1 was used as the immunogen | |
| 0.025 mg | |
| GPCR | |
| 134864 | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction