missing translation for 'onlineSavingsMsg'
Learn More

Trace Amine Receptor 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£167.00 - £335.00

Specifications

Antigen Trace Amine Receptor 1
Concentration 1 mg/mL
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18100562
View Documents
Novus Biologicals
NBP2-24720SS
0.025 mg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18768693
View Documents
Novus Biologicals
NBP2-24720
£335.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Trace Amine Receptor 1 Polyclonal specifically detects Trace Amine Receptor 1 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Trace Amine Receptor 1
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Polyclonal
Purified
RUO
Human, Primate
Q96RJ0
134864
A portion of amino acids 225-250 of human TAAR1 was used as the immunogen
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
GPCR
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1
TAAR1
IgG
Protein G purified
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 86% homologous in mouse and 80% homologous in dog.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.