missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ TR-FRET ER beta Competitive Binding Kit
Description
- The LanthaScreen™ TR-FRET ER beta Competitive Binding Kit provides a sensitive and robust method for high-throughput screening (HTS) of ligands for estrogen receptor (ER) beta. The kit uses a human ER beta ligand-binding domain (LBD) that is tagged with glutathione S-transferase (GST), a terbium-labeled anti-GST antibody, and a fluorescent small-molecule ER ligand (Fluormone™ ES2) in a homogeneous mix-and-read assay format.
- Complete & ready-to-use out of the box - all reagents included, just add your test compounds
- High throughput compatible - TR-FRET assays in a 384-well plate
- Convenient - components available in bulk sizes for larger screening needs
- Includes:
- 1 tube ER beta Ligand Binding Domain Recombinant Protein (store at -68 to -85°C)
- 1 tube of Fluormone ES2 (store at -5 to -30°C)
- 2 bottles NR Buffer (store at -5 to -30°C)
- 1 tube Tb-Anti-GST Ab (store at -5 to -30°C)
- 1 tube of DTT (store at -5 to -30°C)
Specifications
Specifications
| Content And Storage | 1 tube ER beta Ligand Binding Domain Recombinant Protein (store at -68 to -85°C) 1 tube of Fluormone ES2 (store at -5 to -30°C) 2 bottles NR Buffer (store at -5 to -30°C) 1 tube Tb-Anti-GST Ab (store at -5 to -30°C) 1 tube of DTT (store at -5 to -30°C) |
| For Use With (Application) | Competitive Binding Assay, Nuclear Receptor Assay |
| Product Line | LanthaScreen |
| Quantity | 800 x 20 μL assays |
| Type | Competitive Binding Kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?