missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TPPP3 |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16824044
|
Proteintech
15057-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16814044
|
Proteintech
15057-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody detects the 19-20 kDa TPPP3 in HeLa and PC-3 cells. The non-specific bands between 40 and 60 kDa were also occasionally observed.
TPPP3 binds tubulin and has microtubule bundling activity. TPPP3 may play a role in cell proliferation and mitosis.Specifications
| TPPP3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q5PPN5, Q9BW30, Q9CRB6 | |
| 291966, 51673, 67971 | |
| TPPP3 Fusion Protein Ag7040 | |
| Primary | |
| -20°C | |
| TPPP3 |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CGI 38, p20, p25gamma, TPPP/p20, TPPP3 | |
| TPPP3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title