missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15057-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody detects the 19-20 kDa TPPP3 in HeLa and PC-3 cells. The non-specific bands between 40 and 60 kDa were also occasionally observed.
TPPP3 binds tubulin and has microtubule bundling activity. TPPP3 may play a role in cell proliferation and mitosis.Specifications
| TPPP3 | |
| Polyclonal | |
| Unconjugated | |
| TPPP3 | |
| CGI 38, p20, p25gamma, TPPP/p20, TPPP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 291966, 51673, 67971 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5PPN5, Q9BW30, Q9CRB6 | |
| TPPP3 | |
| TPPP3 Fusion Protein Ag7040 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction