missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPD52L2 Mouse anti-Human, Rat, Mouse, Clone: 3B3C6, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TPD52L2 |
|---|---|
| Clone | 3B3C6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17629756
|
Proteintech
68077-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17619756
|
Proteintech
68077-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TPD52L2 Monoclonal antibody apecifically detetects TPD52L2 antigen in Human, Rat, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| TPD52L2 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Rat, Mouse | |
| BC006804 | |
| TPD52L2 | |
| IgG2b | |
| Protein A purification |
| 3B3C6 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Mouse | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| 7165 | |
| fusion protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title