missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPD52L2 Mouse anti-Human, Rat, Mouse, Clone: 3B3C6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68077-1-IG-20UL
This item is not returnable.
View return policy
Description
TPD52L2 Monoclonal antibody apecifically detetects TPD52L2 antigen in Human, Rat, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| TPD52L2 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| TPD52L2 | |
| fusion protein | |
| 20 μL | |
| 7165 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG2b |
| Western Blot, Immunohistochemistry, ELISA | |
| 3B3C6 | |
| Unconjugated | |
| BC006804 | |
| Mouse | |
| Protein A purification | |
| Primary | |
| Human, Rat, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction