missing translation for 'onlineSavingsMsg'
Learn More

TP53INP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17872-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877864

  • £130.00 / 20µL
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

In response to double-strand DNA breaks, promotes p53/TP53 phosphorylation on ′Ser-46′ and subsequent apoptosis.
Specifications

Specifications

TP53INP1
Polyclonal
Unconjugated
TP53INP1
p53DINP1, SIP, Stress induced protein, Teap, TP53DINP1, TP53INP1, TP53INP1A, TP53INP1B
Rabbit
Antigen Affinity Chromatography
RUO
297822, 94241
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q80YE2, Q96A56
TP53INP1
TP53INP1 Fusion Protein Ag12358
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers