missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53INP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | TP53INP1 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877864
|
Proteintech
17872-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867864
|
Proteintech
17872-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
In response to double-strand DNA breaks, promotes p53/TP53 phosphorylation on ′Ser-46′ and subsequent apoptosis.Specifications
TP53INP1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q80YE2, Q96A56 | |
297822, 94241 | |
TP53INP1 Fusion Protein Ag12358 | |
Primary | |
-20°C | |
TP53INP1 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
p53DINP1, SIP, Stress induced protein, Teap, TP53DINP1, TP53INP1, TP53INP1A, TP53INP1B | |
TP53INP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |