missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Test Paper Analysis Case
Shop All Macherey-Nagel Products
Click to view available options
Quantity:
1 Pack
Unit Size:
Each
Description
Simple and fast analysis all in one case
- Fits all strips and papers from the vast Macherey-Nagel test paper and test strip range
- Also fits all the products featured in focus flyers for different applications
Empty case for individual needs
- Can be equipped with almost any test from the Rapid Test product line
- Individual combinations of up to:
- 9 test strips tubes / 5 reagent bottles
- 2 reagent vessels, 4 measuring spoons and 1 beaker
- 2 classics flat boxes of pH-Fix
- 1 test paper reel
- 1 PlopTop tube pH-Fix PT
- 1 test paper box
Specifications
Specifications
| Color | Blue with Black Foam |
| Format | Empty case with foam insert |
| For Use With (Application) | Empty Briefcase Kit |
| For Use With (Equipment) | All products from Rapid Test line |
| Quantity | 1 Pack |
| Size | Small: 28 x 22 x 8cm |
| Type | TP Empty Briefcase Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction