missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toxoplasma Gondii Antibody (TP3cc), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
£393.00
Specifications
| Antigen | Toxoplasma Gondii |
|---|---|
| Clone | TP3cc |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | HRP |
Description
Toxoplasma Gondii Monoclonal antibody specifically detects Toxoplasma Gondii in Protozoa, T. gondii samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Toxoplasma Gondii | |
| Western Blot | |
| HRP | |
| Mouse | |
| Protozoa | |
| t gondii, T. gondii | |
| IgG2a | |
| Protein G purified |
| TP3cc | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Tachyzoites lysate. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title