missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toxoplasma Gondii Antibody (TP3cc), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-2570H
This item is not returnable.
View return policy
Description
Toxoplasma Gondii Monoclonal antibody specifically detects Toxoplasma Gondii in Protozoa, T. gondii samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Toxoplasma Gondii | |
| Monoclonal | |
| HRP | |
| t gondii, T. gondii | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Tachyzoites lysate. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot | |
| TP3cc | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Protozoa | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction