missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethoxy-1,1,3,3-tetramethyldisiloxane, TRC

Product Code. 30010788
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010788 5 g 5g
30134862 1 g 1g
30010452 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010788 Supplier Toronto Research Chemicals Supplier No. TRCD0765155G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18420-09-2
Molecular Formula C8 H22 O3 Si2
Molecular Weight (g/mol) 222.429
Synonym 1,3-Diethoxy-1,1,3,3-tetramethyldisiloxane, 1,1,3,3-Tetramethyl-1,3-diethoxydisiloxane, 1,3-Diethoxytetramethyldisiloxane, α,ω-Diethoxytetramethyldisiloxane;
IUPAC Name ethoxy-[ethoxy(dimethyl)silyl]oxy-dimethylsilane
SMILES CCO[Si](C)(C)O[Si](C)(C)OCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H22O3Si2/c1-7-9-12(3,4)11-13(5,6)10-8-2/h7-8H2,1-6H3
Formula Weight 222.111
Chemical Name or Material 1,3-Diethoxy-1,1,3,3-tetramethyldisiloxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.