CAS RN 18420-09-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,3-Diethoxy-1,1,3,3-tetramethyldisiloxane, TRC
CAS: 18420-09-2 Molecular Formula: C8 H22 O3 Si2 Molecular Weight (g/mol): 222.429 Synonym: 1,3-Diethoxy-1,1,3,3-tetramethyldisiloxane,1,1,3,3-Tetramethyl-1,3-diethoxydisiloxane,1,3-Diethoxytetramethyldisiloxane,α,ω-Diethoxytetramethyldisiloxane; IUPAC Name: ethoxy-[ethoxy(dimethyl)silyl]oxy-dimethylsilane SMILES: CCO[Si](C)(C)O[Si](C)(C)OCC