missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Toremifene, 98+%

Product Code. 15425979
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425979 500 mg 500mg
15405979 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425979 Supplier Thermo Scientific Chemicals Supplier No. J63803.ME
  • ONLINE EXCLUSIVE PRICE
    £89.66
    £70.65 / 500mg
    Save £19.01
    21% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A chlorinated tamoxifen analogue that induces apoptosis in some cells | CAS: 89778-26-7 | C26H28ClNO, C26H28ClNO | 405.97 g/mol

A chlorinated tamoxifen analogue, that induces apoptosis in some cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89778-26-7
Molecular Formula C26H28ClNO
Synonym Toremifene base; 2-(4-[(1Z)-4-Chloro-1,2-diphenyl-1-butenyl]phenoxy)-N,N-dimethylethanamine

Specifications

Description White to Off-white
Quantity 500 mg
Format Powder
Formula Weight 405.96g/mol
Melting Point 108°C to 110°C
Product Type Apoptosis Reagent
For Use With (Application) A chlorinated tamoxifen analogue that induces apoptosis in some cells
Form Powder
Assay ≥98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.