missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TORC2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£186.00 - £408.00
Specifications
| Antigen | TORC2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1.0 - 2.0 ug/ml, Simple Western 1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:20 - 1:100, Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18013634
|
Novus Biologicals
NBP2-22356SS |
0.025 mL |
£186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18778493
|
Novus Biologicals
NBP2-22356 |
0.1 mL |
£408.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TORC2 Polyclonal specifically detects TORC2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TORC2 | |
| Western Blot 1.0 - 2.0 ug/ml, Simple Western 1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:20 - 1:100, Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Hypoxia, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators | |
| PBS with 0.02% Sodium Azide | |
| CREB regulated transcription coactivator 2, CREB-regulated transcription coactivator 2, TORC-2, TORC2transducer of regulated cAMP response element-binding protein (CREB) 2, Transducer of CREB protein 2, Transducer of regulated cAMP response element-binding protein 2 | |
| CRTC2 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Predicted to react in rat and cow based on sequence homology. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q53ET0, Q3U182 | |
| 200186 | |
| A synthetic peptide made to a C-terminal portion of the human Torc2 protein (between residues 600-693) [UniProt Q53ET0] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title