missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TORC2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22356
This item is not returnable.
View return policy
Description
TORC2 Polyclonal specifically detects TORC2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TORC2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CREB regulated transcription coactivator 2, CREB-regulated transcription coactivator 2, TORC-2, TORC2transducer of regulated cAMP response element-binding protein (CREB) 2, Transducer of CREB protein 2, Transducer of regulated cAMP response element-binding protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Predicted to react in rat and cow based on sequence homology. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1.0 - 2.0 ug/ml, Simple Western 1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:20 - 1:100, Immunohistochemistry-Paraffin 1:100 | |
| Q53ET0, Q3U182 | |
| CRTC2 | |
| A synthetic peptide made to a C-terminal portion of the human Torc2 protein (between residues 600-693) [UniProt Q53ET0] | |
| 0.1 mL | |
| Cancer, Hypoxia, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators | |
| 200186 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction