missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOR3A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10647-1-AP-20UL
This item is not returnable.
View return policy
Description
TOR3A (Torsin Family 3 Member A) is a Protein Coding gene. Diseases associated with TOR3A include Apperceptive Agnosia and Speech And Communication Disorders. Gene Ontology (GO) annotations related to this gene include ATPase activity. An important paralog of this gene is TOR1A.Specifications
| TOR3A | |
| Polyclonal | |
| Unconjugated | |
| TOR3A | |
| ADIR, ADIR2, TOR3A, Torsin 3A, Torsin family 3 member A, torsin family 3, member A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 30935, 64222 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9ER38, Q9H497 | |
| TOR3A | |
| TOR3A Fusion Protein Ag1012 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction