missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOR3A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TOR3A |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16817163
|
Proteintech
10647-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16807163
|
Proteintech
10647-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TOR3A (Torsin Family 3 Member A) is a Protein Coding gene. Diseases associated with TOR3A include Apperceptive Agnosia and Speech And Communication Disorders. Gene Ontology (GO) annotations related to this gene include ATPase activity. An important paralog of this gene is TOR1A.Specifications
| TOR3A | |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q9ER38, Q9H497 | |
| 30935, 64222 | |
| TOR3A Fusion Protein Ag1012 | |
| Primary | |
| -20°C | |
| TOR3A |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ADIR, ADIR2, TOR3A, Torsin 3A, Torsin family 3 member A, torsin family 3, member A | |
| TOR3A | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title