missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM22 Antibody (1C9-2), Novus Biologicals™
Mouse Monoclonal Antibody
£271.00
Specifications
| Antigen | TOMM22 |
|---|---|
| Clone | 1C9-2 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Description
TOMM22 Monoclonal specifically detects TOMM22 in Human, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| TOMM22 | |
| 1.0 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones | |
| 1C9-2, hTom22, mitochondrial import receptor subunit TOM22 homolog, mitochondrial import receptor Tom22, MST065, TOM22MSTP065, Translocase of outer membrane 22 kDa subunit homolog, translocase of outer mitochondrial membrane 22 homolog (yeast) | |
| TOMM22 | |
| IgG1 | |
| Protein A or G purified | |
| TOMM22 (1C9-2) |
| 1C9-2 | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Primate | |
| 56993 | |
| Membrane fraction from Vero (monkey kidney-derived) cells. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title