missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM22 Antibody (1C9-2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-10436
This item is not returnable.
View return policy
Description
TOMM22 Monoclonal specifically detects TOMM22 in Human, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| TOMM22 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| TOMM22 | |
| Membrane fraction from Vero (monkey kidney-derived) cells. | |
| 0.05 mL | |
| Membrane Trafficking and Chaperones | |
| 56993 | |
| Human, Primate | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1C9-2 | |
| Unconjugated | |
| 1C9-2, hTom22, mitochondrial import receptor subunit TOM22 homolog, mitochondrial import receptor Tom22, MST065, TOM22MSTP065, Translocase of outer membrane 22 kDa subunit homolog, translocase of outer mitochondrial membrane 22 homolog (yeast) | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| TOMM22 (1C9-2) | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction