missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOM70 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | TOM70 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228174
|
Novus Biologicals
NBP3-33357-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229764
|
Novus Biologicals
NBP3-33357-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TOM70 Monoclonal antibody specifically detects TOM70 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| TOM70 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cellular Markers, Mitochondrial Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 9868 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ90470, KIAA0719, mitochondrial import receptor subunit TOM70, Mitochondrial precursor proteins import receptor, TOM70, Translocase of outer membrane 70 kDa subunit, translocase of outer mitochondrial membrane 70 (yeast) homolog A, translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae), translocase of outer mitochondrial membrane 70 homolog A (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 245-608 of human TOM70 (NP_055635.3).,, Sequence:, KNREPLMPSPQFIKSYFSSFTDDIISQPMLKGEKSDEDKDKEGEALEVKENSGYLKAKQYMEEENYDKIISECSKEIDAEGKYMAEALLLRATFYLLIGNANAAKPDLDKVISLKEANVKLRANALIKRGSMYMQQQQPLLSTQDFNMAADIDPQNADVYHHRGQLKILLDQVEEAVADFDECIRLRPESALAQAQKCFALYRQAYTGNNSSQIQAAMKGFEEVIKKFPRCAEGYALYAQALTDQQQFGKADEMYDKCIDLEPDNATTYVHKGLLQLQWKQDLDRGLELISKAIEIDNKCDFAYETMGTIEVQRGNMEKAIDMFNKAINLAKSEMEMAHLYSLCDAAHAQTEVAKKYGLKPPTL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title