missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOM70 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33357-100ul
This item is not returnable.
View return policy
Description
TOM70 Monoclonal antibody specifically detects TOM70 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TOM70 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| FLJ90470, KIAA0719, mitochondrial import receptor subunit TOM70, Mitochondrial precursor proteins import receptor, TOM70, Translocase of outer membrane 70 kDa subunit, translocase of outer mitochondrial membrane 70 (yeast) homolog A, translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae), translocase of outer mitochondrial membrane 70 homolog A (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 245-608 of human TOM70 (NP_055635.3).,, Sequence:, KNREPLMPSPQFIKSYFSSFTDDIISQPMLKGEKSDEDKDKEGEALEVKENSGYLKAKQYMEEENYDKIISECSKEIDAEGKYMAEALLLRATFYLLIGNANAAKPDLDKVISLKEANVKLRANALIKRGSMYMQQQQPLLSTQDFNMAADIDPQNADVYHHRGQLKILLDQVEEAVADFDECIRLRPESALAQAQKCFALYRQAYTGNNSSQIQAAMKGFEEVIKKFPRCAEGYALYAQALTDQQQFGKADEMYDKCIDLEPDNATTYVHKGLLQLQWKQDLDRGLELISKAIEIDNKCDFAYETMGTIEVQRGNMEKAIDMFNKAINLAKSEMEMAHLYSLCDAAHAQTEVAKKYGLKPPTL | |
| 100 μL | |
| Apoptosis, Cellular Markers, Mitochondrial Markers, Signal Transduction | |
| 9868 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction