missing translation for 'onlineSavingsMsg'
Learn More

Tolterodine L-Tartrate, TRC

Product Code. 30133170
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133170 100 mg 100mg
30133093 25 mg 25mg
30010426 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133170 Supplier Toronto Research Chemicals Supplier No. TRCT535800100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124937-52-6
Molecular Formula C22 H31 N O . C4 H6 O6
Molecular Weight (g/mol) 475.57
Synonym Tolterodine Tartrate, 2-[(1R)-3-[Bis(1-methylethyl)amino]-1-phenylpropyl]-4-methylphenol (2R,3R)-2,3-dihydroxybutanedioate (1:1), (R)-2-[3-[Bis(1-methylethyl)amino]-1-phenylpropyl]-4-methylphenol [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), Phenol, 2-[(1R)-3-[bis(1-methylethyl)amino]-1-phenylpropyl]-4-methyl-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), Phenol, 2-[(1R)-3-[bis(1-methylethyl)amino]-1-phenylpropyl]-4-methyl-, (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt) (9CI), Phenol, 2-[3-[bis(1-methylethyl)amino]-1-phenylpropyl]-4-methyl-, (R)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), (R)-(+)-N,N-Diisopropyl-3-(2-hydroxy-5-methylphenyl)-3-phenylpropylamine L-tartrate, (R)-Tolterodine L-tartrate, (R)-Tolterodine tartrate, Detrol, Detrusitol, PNU 200583E, Terol, Tolterodine L-tartrate, Tolterodine tartaric acid, Tolterodine tartrate, Tolterodinium (+)-(2R,3R)-hydrogen tartrate (1:1)
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;2-[(1R)-3-[di(propan-2-yl)amino]-1-phenylpropyl]-4-methylphenol
SMILES CC(C)N(CC[C@H](c1ccccc1)c2cc(C)ccc2O)C(C)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H31NO.C4H6O6/c1-16(2)23(17(3)4)14-13-20(19-9-7-6-8-10-19)21-15-18(5)11-12-22(21)24;5-1(3(7)8)2(6)4(9)10/h6-12,15-17,20,24H,13-14H2,1-5H3;1-2,5-6H,(H,7,8)(H,9,10)/t20-;1-,2-/m11/s1
Formula Weight 475.257
Chemical Name or Material Tolterodine tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.