CAS RN 124937-52-6
CAS RN 124937-52-6
IUPAC Name:
(2R,3R)-2,3-dihydroxybutanedioic acid; 2-[(1S)-3-[bis(propan-2-yl)amino]-1-phenylpropyl]-4-methylphenol
Synonyms:
L(+)-tartaric acid; tolterodine
Molecular Weight (g/mol):
475.58
Molecular Formula:
C26H37NO7
InChi Key:
TWHNMSJGYKMTRB-CDHDNNKHSA-N
SMILES:
O[C@H]([C@@H](O)C(O)=O)C(O)=O.CC(C)N(CC[C@@H](C1=CC=CC=C1)C1=CC(C)=CC=C1O)C(C)C
1
–
3
of
3
results
1
Tolterodine Tartrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Tolterodine Tartrate, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
3
Tolterodine tartrate, MedChemExpress
MedChemExpress Tolterodine Tartrate (Kabi-2234; PNU-200583E) is a potent muscarinic receptor antagonist and shows selectivity for the urinary bladder over salivary glands in vivo.