missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOE1 Recombinant Rabbit Monoclonal Antibody (24GB1860)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen TOE1
Clone 24GB1860
Concentration 2.5 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276469
GREENER_CHOICE
View Documents
Invitrogen™
MA557790
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276014
View Documents
Invitrogen™
MA557791
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TOE1 Recombinant Monoclonal Antibody for Western Blot, Flow

Inhibits cell growth rate and cell cycle. Induces CDKN1A expression as well as TGF-beta expression. Mediates the inhibitory growth effect of EGR1. Involved in the maturation of snRNAs and snRNA 3'-tail processing (PubMed:28092684). [UniProt]
Specifications

Specifications

TOE1
2.5 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q96GM8, Q9D2E2
114034, 298443, 68276
A synthesized peptide derived from human TOE1 (aa 15-77).
Antibody
24GB1860
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
TOE1
4930584N22Rik; 4933424D16Rik; AI413517; hCaf1z; Target of EGR1 protein 1; target of EGR1, member 1 (nuclear); TOE1
TOE1
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.