missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOE1 Recombinant Rabbit Monoclonal Antibody (24GB1860)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557791
This item is not returnable.
View return policy
Description
TOE1 Recombinant Monoclonal Antibody for Western Blot, Flow
Inhibits cell growth rate and cell cycle. Induces CDKN1A expression as well as TGF-beta expression. Mediates the inhibitory growth effect of EGR1. Involved in the maturation of snRNAs and snRNA 3'-tail processing (PubMed:28092684). [UniProt].
Specifications
| TOE1 | |
| Recombinant Monoclonal | |
| 2.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96GM8, Q9D2E2 | |
| TOE1 | |
| A synthesized peptide derived from human TOE1 (aa 15-77). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1860 | |
| Unconjugated | |
| TOE1 | |
| 4930584N22Rik; 4933424D16Rik; AI413517; hCaf1z; Target of EGR1 protein 1; target of EGR1, member 1 (nuclear); TOE1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114034, 298443, 68276 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto