Learn More
Invitrogen™ TNK2 (ACK) Recomibinant Human Protein
TNK2 (ACK) (activated Cdc42-associated kinase) is a non-receptor tyrosine kinase that signals downstream of Cdc42.
Brand: Invitrogen™ PV4808
Code : NEW
Additional Details : Weight : 0.01000kg
Description
TNK2 is known to be involved in tumorigenesis and metastasis, particularly in prostate cancer.
Specifications
NP_005772.3 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
TNK2 (ACK) | |
Human | |
GST Tag | |
Frozen |
Nuclear Receptor Assay | |
69.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
ACK Kinase Family | |
Recombinant | |
Baculovirus |