missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNK2 (ACK) Recomibinant Human Protein
TNK2 (ACK) (activated Cdc42-associated kinase) is a non-receptor tyrosine kinase that signals downstream of Cdc42.
£1602.00 - £6900.00
Specifications
Accession Number | NP_005772.3 |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 69.3kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15498683
|
Invitrogen™
PR8863A |
100μg |
£1602.00
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15428763
|
Invitrogen™
PV4808 |
1mg |
£6900.00
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TNK2 is known to be involved in tumorigenesis and metastasis, particularly in prostate cancer.
Specifications
NP_005772.3 | |
69.3kDa | |
RUO - research use only | |
TNK2 (ACK) | |
Human | |
GST Tag | |
Frozen |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
ACK Kinase Family | |
Recombinant | |
Baculovirus |