missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tnk1 Antibody (1B5G3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47379-0.025ml
This item is not returnable.
View return policy
Description
Tnk1 Monoclonal specifically detects Tnk1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Tnk1 | |
| Monoclonal | |
| Unconjugated | |
| CD38 negative kinase 1, EC 2.7.10, EC 2.7.10.2, MGC46193, non-receptor tyrosine-protein kinase TNK1, tyrosine kinase, non-receptor, 1 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Western Blot | |
| 1B5G3 | |
| Western Blot 1: 500 - 1:2000, ELISA 1:10000 | |
| TNK1 | |
| Purified recombinant fragment of TNK1(aa451-560) expressed in E. Coli. | |
| 0.025 mL | |
| Protein Kinase | |
| 8711 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction