missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tnk1 Antibody (1B5G3), Novus Biologicals™
Mouse Monoclonal Antibody
£135.00
Specifications
| Antigen | Tnk1 |
|---|---|
| Clone | 1B5G3 |
| Dilution | Western Blot 1: 500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Tnk1 Monoclonal specifically detects Tnk1 in Human samples. It is validated for Western Blot, ELISA.Specifications
| Tnk1 | |
| Western Blot 1: 500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 8711 | |
| Purified recombinant fragment of TNK1(aa451-560) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1B5G3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| CD38 negative kinase 1, EC 2.7.10, EC 2.7.10.2, MGC46193, non-receptor tyrosine-protein kinase TNK1, tyrosine kinase, non-receptor, 1 | |
| TNK1 | |
| IgG1 | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title