missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF RII/TNFRSF1B Rabbit anti-Human, Clone: R00N6, Novus Biologicals™
Rabbit Monoclonal Antibody
£411.00 - £549.00
Specifications
| Antigen | TNF RII/TNFRSF1B |
|---|---|
| Clone | R00N6 |
| Applications | Neutralization |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18267047
|
Novus Biologicals
NBP2-89487-200UG |
200 μg |
£411.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18239017
|
Novus Biologicals
NBP2-89487-500UG |
500 μg |
£549.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Human samples. It is validated for Neutralization.Specifications
| TNF RII/TNFRSF1B | |
| Neutralization | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Signal Transduction, Zinc Finger | |
| 0.2 um filtered solution in Histidine and Arginine buffer, 120mM NaCl, 0.02% Tween 80, pH6.0 | |
| 7133 | |
| IgG | |
| Protein A purified |
| R00N6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
| Recombinant Human TNF RII/TNFRSF1B protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title