missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF RII/TNFRSF1B Rabbit anti-Human, Clone: R00N6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89487-500UG
This item is not returnable.
View return policy
Description
TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Human samples. It is validated for Neutralization.
Specifications
| TNF RII/TNFRSF1B | |
| Monoclonal | |
| Unconjugated | |
| CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
| Recombinant Human TNF RII/TNFRSF1B protein | |
| 500 μg | |
| Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Signal Transduction, Zinc Finger | |
| 7133 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Neutralization | |
| R00N6 | |
| 0.2 um filtered solution in Histidine and Arginine buffer, 120mM NaCl, 0.02% Tween 80, pH6.0 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction