missing translation for 'onlineSavingsMsg'
Learn More

Mouse TNF-alpha Antibody, R&D Systems™
GREENER_CHOICE

Hamster Monoclonal Antibody has been used in 3 publications

£168.00 - £1530.00

Specifications

Antigen TNF-alpha
Clone TN3-19.12
Dilution Western Blot 1 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18768438
GREENER_CHOICE
View Documents
R&D Systems
MAB410-SP
25 μg
£168.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18747669
GREENER_CHOICE
View Documents
R&D Systems
MAB410-100
100 μg
£387.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18710978
GREENER_CHOICE
View Documents
R&D Systems
MAB410-500
500 μg
£1530.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TNF-alpha Monoclonal specifically detects TNF-alpha in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

TNF-alpha
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
7124
E. coli-derived recombinant mouse TNF-alpha
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects recombinant mouse (rm)TNF-alpha in direct ELISAs and Western blots. Shows 50% cross-reactivity with rrTNF-alpha and no cross-reactivity with 17 other tested proteins.
TN3-19.12
Western Blot
Unconjugated
Hamster
Mouse
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
TNF
IgG
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.