missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TNF-alpha Antibody, R&D Systems™

Hamster Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB410-SP
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Mouse samples. It is validated for Western Blot.
Specifications
| TNF-alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TNF | |
| E. coli-derived recombinant mouse TNF-alpha | |
| 25 μg | |
| Primary | |
| Detects recombinant mouse (rm)TNF-alpha in direct ELISAs and Western blots. Shows 50% cross-reactivity with rrTNF-alpha and no cross-reactivity with 17 other tested proteins. | |
| Mouse | |
| Purified |
| Western Blot | |
| TN3-19.12 | |
| Western Blot 1 ug/mL | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Hamster | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7124 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction