missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMTsixplex™ Isobaric Label Reagent Set

Product Code. 15680079
Change view
Click to view available options
:
16 x 0.2mg/TMT label
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15680079 16 x 0.2mg/TMT label
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15680079 Supplier Thermo Scientific™ Supplier No. 90308

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TMT™ Isobaric Mass Tagging Kits and Reagents enable multiplex quantitation of proteins extracted from cells and tissues using tandem mass spectrometry.

Features:
  • Powerful - Concurrent MS analysis of multiple samples increases sample throughput and enables relative quantitation of up to six different samples derived from cells, tissues or biological fluids
  • Consistent - Identical reagent structure and performance among TMTzero™, TMTduplex™, TMTsixplex™ and TMT10plex™ reagents allows efficient transition from method development to multiplex quantitation, enabling biomarker discovery research across platforms and datasets
  • Robust - Consistent chemistry allows efficient transition from method development to multiplex quantitation, enabling biomarker discovery research
  • Efficient - Amine-reactive NHS-ester activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
  • Compatible - Optimized for use with high resolution Thermo Scientific MS/MS platforms, such as Q Exactive, Orbitrap Elite™ and Orbitrap Fusion™ Tribrid™ instruments with data analysis fully supported by Proteome Discoverer™ 1.0 and above
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein quantitation in Mass spectrometry
Kit Contents 16 x 0.2 mg each of TMT6-126 Label Reagent, TMT6-127 Label Reagent, TMT6-128 Label Reagent, TMT6-129 Label Reagent, TMT6-130 Label Reagent, TMT6-131 Label Reagent
Sufficient For 96 reactions (16 x 6-plex experiments)
Content And Storage Freezer (-5°C to -30°C)
Product Type Isobaric Label Reagent Set

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.