missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMTpro™ 32plex and TMTpro™ Deuterated Label Reagents

Product Code. 30193507 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
134C and 135CD Label Reagents
135CD Label Reagent
16plex Deuterated Label Reagent Set
32plex Label Reagent Matched Set
Quantity:
1 x 0.5 mg (per tag)
1 x 5 mg
1 x 5 mg (per tag)
Unit Size:
0.50mg
5mg
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity unitSize
30193507 16plex Deuterated Label Reagent Set 1 x 5 mg (per tag) Each
30197496 32plex Label Reagent Matched Set 1 x 5 mg (per tag) Each
30193506 134C and 135CD Label Reagents 1 x 5 mg (per tag) Each
30193508 135CD Label Reagent 1 x 5 mg 5mg
31759102 16plex Deuterated Label Reagent Set 1 x 0.5 mg (per tag) 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30193507 Supplier Thermo Scientific™ Supplier No. A40000817
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

TMTpro deuterated label reagents enable multiplexing of up to 35 samples, allowing greater throughput for protein identification and quantitation using tandem mass spectrometry.

Thermo Scientific TMTpro label reagents are the next generation of tandem mass tags, designed to increase the level of sample multiplexing without compromising on protein identification and quantitation. TMTpro deuterated label reagents enable multiplexing of up to 35 samples, for even greater throughput for protein identification and quantitative analysis by tandem mass spectrometry (MS/MS).

Features of the TMTpro 32plex Label Reagent Set include:

  • Multiplex—concurrent MS analysis of up to 35 samples derived from cells, tissues, or biological fluids
  • Robust—increased multiplex capability results in fewer missing quantitative values among samples and enables confidence among replicates
  • Efficient—amine-reactive, NHS ester-activated reagents help ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
  • Optimized for high-resolution MS/MS analysis using Orbitrap platforms with data processing by Proteome Discoverer 3.2

After fragmentation, each TMTpro tag generates a reporter ion with unique mass (e.g., 126-135 Da) in the low-mass region of the high-resolution MS/MS spectrum that is used for multiplexed relative quantitation of protein abundance.

Advantages of TMTpro tags

The advantages of TMTpro tags include increased multiplexed relative quantitation, increased sample throughput, and fewer missing quantitative values among samples. With these capabilities, the TMTpro label reagents are suited for applications requiring higher multiplexing and fewer missing values, such as inhibitor dose response experiments, time course experiments, thermal shift assays, or single-cell analysis. When combined with the industry-leading high-resolution Orbitrap instruments and software, TMTpro reagents provide integrated total solutions for quantitative proteomic analysis.

Combining TMTpro non-deuterated label reagents with their deuterated counterparts allows multiplexing up to 35 samples. Control channels within each set (non-deuterated and deuterated) are necessary to compare data across sub-plex. TMTpro-134C and TMTpro-135CD reagents are offered for use as control channels in combination with the 32plex set. TMTpro-135CD reagent is offered separately to combine with TMTpro 18plex and TMTpro 16plex deuterated label reagents to achieve 35-sample multiplexing, including multiple controls for data analysis.

For 32plex experiments, use TMTpro 32plex Label Reagent Matched Set (Cat. No. A40000839) or TMTpro 16plex Deuterated Label Reagent Set (Cat. No. A40000817) and TMTpro 16plex Label Reagent Set (Cat. No. A44520).

For 34plex experiments use TMTpro 32plex Label Reagent Matched Set (Cat. No. A40000839) and TMTpro-134C and TMTpro-135CD Label Reagents (Cat. No. A40000853) or TMTpro 16plex Deuterated Label Reagent Set (Cat. No. A40000817), TMTpro 16plex Label Reagent Set (Cat. No. A44520), and TMTpro-134C and TMTpro-135CD Label Reagents (Cat. No. A40000853).

For 35plex experiments use TMTpro 16plex Deuterated Label Reagent Set (Cat. No. A40000817), TMTpro-135CD Label Reagent (Cat. No. A40000818), and TMTpro 18plex Label Reagent Set (Cat. No. A52045).

Related products

For high resolution analysis of TMTpro-labeled peptides, the recommended LC column for the Nanospray Flex source is the Acclaim PepMap 100 C18 LC Column (Cat. No. 164942, 164939). For the EASY-Spray source, the recommended LC column is the EASY-Spray PepMap Neo UHPLC Column (Cat. No. ES75500PN) or EASY-Spray HPLC Column (Cat. No. ES902 or ES903).

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type 16plex Deuterated Label Reagent Set
Product Line TMTpro
Content And Storage • TMTpro-127D Label Reagent, 1 x 5 mg
• TMTpro-128ND Label Reagent, 1 x 5 mg
• TMTpro-128CD Label Reagent, 1 x 5 mg
• TMTpro-129ND Label Reagent, 1 x 5 mg
• TMTpro-129CD Label Reagent, 1 x 5 mg
• TMTpro-130ND Label Reagent, 1 x 5 mg
• TMTpro-130CD Label Reagent, 1 x 5 mg
• TMTpro-131ND Label Reagent, 1 x 5 mg
• TMTpro-131CD Label Reagent, 1 x 5 mg
• TMTpro-132ND Label Reagent, 1 x 5 mg
• TMTpro-132CD Label Reagent, 1 x 5 mg
• TMTpro-133ND Label Reagent, 1 x 5 mg
• TMTpro-133CD Label Reagent, 1 x 5 mg
• TMTpro-134ND Label Reagent, 1 x 5 mg
• TMTpro-134CD Label Reagent, 1 x 5 mg
• TMTpro-135ND Label Reagent, 1 x 5 mg

Store below -20°C.
Label or Dye Tandem Mass Tag™ (TMT) Labels
Shipping Condition Dry Ice
Starting Material Cell Lysate
Final Product Type Peptide
Detection Method Mass Spectrometry
Workflow Step Peptide Labeling and Quantitation
Quantity 1 x 5 mg (per tag)
Form Lyophilized Powder
Show More Show Less
What is the shelf life for the TMTpro 32plex and TMTpro Deuterated Label Reagents?

The TMTpro 32plex and TMTpro Deuterated Label Reagents are guaranteed for one year from the date of receipt when stored at the recommended temperature in original, unopened packaging.

If I already have TMTpro 16plex Label Reagent Set (Cat. No. A44520), what do I need to purchase to perform 32plex experiments?

You would need to use TMTpro 16plex Label Reagent Set (Cat. No. A44520) in combination with TMTpro 16plex Deuterated Label Reagent Set (Cat. No. A40000817).

Can I purchase the TMTpro-134C and TMTpro-135CD Label Reagents separately from the TMTpro 32plex and TMTpro Deuterated Label Reagents?

Yes, 134C and 135CD Label Reagents (Cat. No. A40000853) provides TMTpro-134C and TMTpro-135CD tags in two separate vials with each vial containing 5 mg of the tag in lyophilized format. 135CD Label Reagent (Cat. No. A40000818) provides one vial containing 5 mg of TMTpro-135 CD tag in lyophilized format.

What is provided with the 134C and 135CD Label Reagents (Cat. No. A40000853)?

134C and 135CD Label Reagents (Cat. No. A40000853) provides two TMTpro tags, TMTpro-134C and TMTpro-135CD. The tags are supplied in two separate vials, with each vial containing 5 mg of the tag in lyophilized format.

Do you offer stand-alone replacement tags for TMTpro 32plex and TMTpro Deuterated Label Reagents?

TMTpro 135CD is the only tag offered as a standalone product (Cat. No. A40000818, 135CD Label Reagent).

What are the different size formats that the TMTpro 32plex and TMTpro Deuterated Label Reagents are available in?

The TMTpro 32plex and TMTpro Deuterated Label Reagents can be purchased in four different size formats:
- Cat. No. A40000817: 16plex Deuterated Label Reagent Set, 1 x 5 mg (per tag)
- Cat. No. A40000839: 32plex Label Reagent Matched Set, 1 x 5 mg (per tag); Note that this is a combo kit of TMTpro 16plex Label Reagent Set (Cat. No. A44520) and TMTpro 16plex Deuterated Label Reagent Set (Cat. No. A40000817)
- Cat. No. A40000853: 134C and 135CD Label Reagents, 1 x 5 mg (per tag)
- Cat. No. A40000818: 135CD Label Reagent, 1 x 5 mg (per tag)

Can the TMT and TMTpro reagents be mixed?

No, the TMT and TMTpro reagents cannot be mixed because the reporter ions are in the same region.

Can TMTpro label reagents dissolved in anhydrous acetonitrile be dried down and stored for later use?

Yes, unused TMTpro label reagent that has been dissolved in anhydrous acetonitrile can be aliquoted into tubes, dried under vacuum at 25-30 degrees C, purged with nitrogen, and stored in a sealed bag with desiccant at -20 degrees C.

Can I combine TMTpro tags with TMT tags?

We do not recommend mixing TMTpro and TMT tags, as they differ in chemical structure.

At which step should I label my mass spec samples with TMT reagents?

TMT labeling can be performed after reduction, alkylation, digestion, or prior to cleanup, if the sample is buffered at appropriate pH (8-8.5) in a suitable buffer without primary amines (e.g., Tris, glycine). TMT labeling can also be performed after peptide clean-up. Labeling of peptides after clean-up enables measuring and normalizing of the peptide samples for equal mixing.

Do I have to apply correction factors for analyzing data for TMT/TMTpro-labeled samples?

Applying correction factors is required for more accurate relative protein quantitation as the isotopic impurity is variable among the tags, ranging from 5-10%.

In the EasyPep sample preparation workflow, it is mentioned that TMT/TMTpro reagent can be added to peptides before or after the clean-up step. Are there any differences between these two methods, regarding the labeling efficiency or relative quantitation?

The EasyPep kit chemistry is fully compatible with TMT/TMTpro reagents. Most commonly, TMT/TMTpro reagents are used to label peptides immediately after digestion and before peptide cleanup. This enables combining samples for a single cleanup step which reduces variability for more reproducible quantitative measurements. Another option is to label samples after peptide cleanup. This approach provides efficient labeling of the samples but may require the use of a peptide quantification assay to ensure that samples are equally mixed before LC-MS analysis. Both the workflows should provide high labelling efficiencies using our recommended TMT/TMTpro reagent to sample ratios.

I have low levels (<95%) of TMT-labeling for my samples. What should I check?

Labeling efficiency can be determined for individual labeled samples by searching for peptides using TMT/TMTpro as a variable (i.e., dynamic) modification peptide amino terminus and lysines. The ratio of peptide to tag by mass (w:w) should be 1:4 to 1:8 for complete labeling of most samples.

Low labeling efficiency can also be caused by improper storing or handling of TMT/TMTpro reagents that leads to hydrolysis of -NHS groups on the tag, rendering the reagents to be less reactive.

Can I use TMT reagents in combination with TMTpro reagents?

TMT reagents and TMTpro reagents are different chemicals with different masses. Therefore, we do not recommend mixing the two tags as the combined samples would be significantly more complex resulting in fewer quantified proteins.

What sample types can I use with different mass spec quantitation systems?

Label-free, TMT reagents, and TMTpro reagents can be used with any protein sample type. SILAC and Neucode quantitation can only be used with cell lines and model organisms that can be metabolically labeled.

How many samples can I combine for simultaneous, multiplex proteomic mass spec quanitification?

SILAC can be used to multiplex 2 to 3 samples. TMT reagents can be used to multiplex 2, 6, 10, and 11 samples. TMTpro reagents can be used to measure up to 16-18 samples simultaneously. 32 samples can be measured using a combination of TMTpro non-deuterated and deuterated tags.

What is the main purpose of Tandem Mass Tag (TMT) reagents?

Tandem Mass Tag technology is used to individually label different protein samples so they can be combined into a single sample for LC-MS analysis. The major advantages of this workflow are higher sample throughput, less instrument analysis time, high precision of peptide quantitation among replicates, and fewer missing quantified proteins among different samples.

Is TMT quantitation relative or absolute?

TMT quantitation is relative between samples unless a heavy peptide standard is included for comparison.

What is the main purpose of TMT (Tandem Mass Tag) reagents?

TMT technology is used to perform multiplex quantitation of proteins extracted from cells and tissues. Rather than quantify single proteins (for example with an antibody), here, large numbers of proteins can be quantified in a single run.

When should I use quadrupole versus ion trap isolation?

Quadrupole isolation is faster because ions must travel through the quadrupole analyzer regardless of the analysis type chosen. As a result, there is little to no time penalty when using the quadrupole for MS2 isolation. In such a case, quadrupole transmission efficiency will depend on isolation width, m/z, and the cleanliness of the analyzer. Ion trap isolation is best suited for MS3 experiments, including those performed using multinotch isolation for TMT analysis; these types of isolation events are not possible with the quadrupole analyzer.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.