missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMT10plex™ Isobaric Label Reagents and Kits

Product Code. 15698065 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 x 0.8 mg (per tag), 10 reactions
1 x 5 mg (per tag), 60 reactions
3 x 0.8 mg
3 x 0.8 mg (per tag), 30 reactions
8 x 0.2 mg
Product Type:
Isobaric Label Reagent Set
Isobaric Mass Tag Labeling Kit
Unit Size:
10 reactions
30 reactions
60 reactions
80 reactions
This item is not returnable. View return policy

Product Code. 15698065

Brand: Thermo Scientific™ 90309

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TMT10plex Isobaric Label Reagents and Kits contain ten isobaric compounds with the same mass and chemical structure (i.e., isotopomeric) composed of an amine-reactive NHS-ester group, a spacer arm, and a mass reporter.

TMT10plex Isobaric Label Reagents and Kits contain ten isobaric compounds with the same mass and chemical structure (i.e., isotopomeric) composed of an amine-reactive NHS-ester group, a spacer arm, and a mass reporter. Up to ten different peptide samples prepared from cells or tissues can be labeled in parallel and then combined for analysis. For each sample, a unique reporter mass (i.e., TMT10 126-131 Da) in the low-mass region of the high-resolution MS/MS spectrum is used to measure relative protein expression levels during peptide fragmentation and tandem mass spectrometry.

The Tandem Mass Tag (TMT) Isobaric Label reagents and kits are specifically designed to enable identification and quantitation of proteins in different samples using tandem mass spectrometry. The TMT10plex label reagents share an identical structure with TMTzero, TMTduplex, and TMTsixplex reagents, but contain different numbers and combinations of 13C and 15N isotopes in the mass reporter. The different isotopes result in a 10-plex set of tags that have mass differences in the reporter that can be detected using high resolution Orbitrap MS instruments.

Advantages of the TMT10plex label reagents include increased multiplex relative quantitation, increased sample throughput, and fewer missing quantitative values among samples. TMT10plex label reagents are ideal for analysis of multiple protein samples from inhibitor dose response experiments, time course experiments, or improving statistics on biological replicates.

Features of TMT10plex Isobaric Label reagent sets and mass tagging kits
Powerful—multiplexed MS analysis increases sample throughput and enables relative quantitation of up to ten different samples derived from cells, tissues, or biological fluids
Consistent—identical reagent structure and performance among TMTzero, TMTduplex, TMTsixplex, and TMT10plex reagents allows efficient transition from method development to multiplex quantitation, enabling biomarker discovery research across platforms and datasets
Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
Efficient—amine-reactive, NHS-ester-activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
Compatible—optimized for use with high resolution Orbitrap MS/MS platforms, with data analysis fully supported by Proteome Discoverer 2.3.

Applications:
• Protein identification and quantitation from multiple samples of cells, tissue, or biological fluids
• Protein expression profiling of normal vs. disease states or control vs. treated
• Quantitative analysis of proteins for which no antibodies are available
• Identification and quantitation of membrane and post-translationally modified proteins
• Identification and quantification of hundreds to thousands of proteins in a single experiment

TMT Reagents are provided as standalone sets or in optimized kit formats containing all necessary reagents and controls for maximum flexibility, convenience, and reliability. When combined with the industry-leading, high resolution Thermo Scientific Orbitrap instruments and software, TMT10plex reagents provide integrated total solutions for quantitative protein expression analysis.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Isobaric Label Reagent Set
Product Line TMT10plex
Content And Storage • 8 x 0.2 mg TMT10–126 Label Reagent
• 8 x 0.2 mg TMT10–127N Label Reagent
• 8 x 0.2 mg TMT10–127C Label Reagent
• 8 x 0.2 mg TMT10–128N Label Reagent
• 8 x 0.2 mg TMT10–128C Label Reagent
• 8 x 0.2 mg TMT10–129N Label Reagent
• 8 x 0.2 mg TMT10–129C Label Reagent
• 8 x 0.2 mg TMT10–130N Label Reagent
• 8 x 0.2 mg TMT10–130C Label Reagent
• 8 x 0.2 mg TMT10–131 Label Reagent

Store in freezer (–5 to –30°C).

Label or Dye Tandem Mass Tag™ (TMT) Labels
Shipping Condition Dry Ice
Starting Material Peptides
Detection Method Mass Spectrometry
Workflow Step Peptide quantitation
Quantity 8 x 0.2 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.