missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress TMRE

Product Code. 16438778
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16438778

Brand: MedChemExpress HYD0985A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TMRE is a mitochondria specific dye (λex=550 nm, λem=575 nm).

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Product Type Dye Reagents
Quantity 10 mg
Shipping Condition Room temperature
Solubility DMSO : 27.78 mg/mL (53.95 mM; Need ultrasonic)
Target HBV
Molecular Weight (g/mol) 514.95 g/mol
CAS 115532-52-0
Synonym Tetramethylrhodamine ethyl ester perchlorate
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.