missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM9B Antibody (S02-9C6), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | TMEM9B |
|---|---|
| Clone | S02-9C6 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636153
|
Novus Biologicals
NBP3-19790-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686353
|
Novus Biologicals
NBP3-19790-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM9B Monoclonal antibody specifically detects TMEM9B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Sandwich ELISASpecifications
| TMEM9B | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C11orf15, chromosome 11 open reading frame 15, TMEM9 domain family, member B, transmembrane protein 9B | |
| Recombinant protein of human TMEM9B (Uniprot # Q9NQ34) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S02-9C6 | |
| Western Blot, Flow Cytometry, ELISA, Sandwich ELISA | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 56674 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title