missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM9B Antibody (S02-9C6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19790-50ul
This item is not returnable.
View return policy
Description
TMEM9B Monoclonal antibody specifically detects TMEM9B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Sandwich ELISA
Specifications
| TMEM9B | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Sandwich ELISA | |
| S02-9C6 | |
| Unconjugated | |
| C11orf15, chromosome 11 open reading frame 15, TMEM9 domain family, member B, transmembrane protein 9B | |
| Recombinant protein of human TMEM9B (Uniprot # Q9NQ34) | |
| 50 μg | |
| Signal Transduction | |
| 56674 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction