missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM67 Recombinant Rabbit Monoclonal Antibody (23GB4990), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | TMEM67 |
|---|---|
| Clone | 23GB4990 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088878
![]() |
Invitrogen™
MA552896 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088874
|
Invitrogen™
MA552897 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM67 Recombinant Monoclonal antibody specifically detects TMEM67 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| TMEM67 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P0C152, Q5HYA8, Q8BR76 | |
| 313067, 329795, 91147 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4990 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 5330408M12Rik; B230117O07; b2b1163.1Clo; b2b1291.1Clo; JBTS6; meckel syndrome type 3 protein; Meckel syndrome type 3 protein homolog; Meckelin; Mks3; NPHP11; TMEM67; TNEM67; transmembrane protein 67 | |
| A synthesized peptide derived from human Meckelin (801-880AA). | |
| Primary | |
| -20°C | |
| TMEM67 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title