missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM67 Recombinant Rabbit Monoclonal Antibody (23GB4990), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552896
This item is not returnable.
View return policy
Description
The protein encoded by this gene localizes to the primary cilium and to the plasma membrane. The gene functions in centriole migration to the apical membrane and formation of the primary cilium. Multiple transcript variants encoding different isoforms have been found for this gene. Defects in this gene are a cause of Meckel syndrome type 3 (MKS3) and Joubert syndrome type 6 (JBTS6).
Specifications
| TMEM67 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TMEM67 | |
| 5330408M12Rik; B230117O07; b2b1163.1Clo; b2b1291.1Clo; JBTS6; meckel syndrome type 3 protein; Meckel syndrome type 3 protein homolog; Meckelin; Mks3; NPHP11; TMEM67; TNEM67; transmembrane protein 67 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 313067, 329795, 91147 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4990 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P0C152, Q5HYA8, Q8BR76 | |
| TMEM67 | |
| A synthesized peptide derived from human Meckelin (801-880AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction