missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM43 Recombinant Rabbit Monoclonal Antibody (24GB2325)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | TMEM43 |
|---|---|
| Clone | 24GB2325 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276027
![]() |
Invitrogen™
MA557796 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275219
|
Invitrogen™
MA557797 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM43 Recombinant Monoclonal Antibody for Western Blot, Flow
This gene belongs to the TMEM43 family. Defects in this gene are the cause of familial arrhythmogenic right ventricular dysplasia type 5 (ARVD5), also known as arrhythmogenic right ventricular cardiomyopathy type 5 (ARVC5). Arrhythmogenic right ventricular dysplasia is an inherited disorder, often involving both ventricles, and is characterized by ventricular tachycardia, heart failure, sudden cardiac death, and fibrofatty replacement of cardiomyocytes. This gene contains a response element for PPAR gamma (an adipogenic transcription factor), which may explain the fibrofatty replacement of the myocardium, a characteristic pathological finding in ARVC.Specifications
| TMEM43 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| TMEM43 | |
| 1200015A22Rik; ARVC5; ARVD5; EDMD7; LUMA; Protein LUMA; Tmem43; Transmembrane protein 43; UNQ2564/PRO6244 | |
| TMEM43 | |
| Primary | |
| Affinity chromatography |
| 24GB2325 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5XIP9, Q9BTV4, Q9DBS1 | |
| 362401, 74122, 79188 | |
| A synthesized peptide derived from human TMEM43 (aa 100-135). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title