missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM27 Monoclonal Antibody (C5), Invitrogen™

Mouse Monoclonal Antibody
£178.00 - £386.00
Specifications
| Antigen | TMEM27 |
|---|---|
| Clone | C5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17779883
|
Invitrogen™
MA543559 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17644016
![]() |
Invitrogen™
MA543558 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM27 (Transmembrane protein 27) is also named as collectrin and Belongs to the TMEM27 family. It is a stimulator of beta cell replication and a membrane protein cleaved and shed by pancreatic beta cells that has been proposed as a beta cell mass biomarker. Western blot analysis of mouse collecting duct cell lines detected Tmem27 at an apparent molecular mass of 32 kDa. TMEM27 is an N-glycosylated protein and forms dimers, with MW of 64-75 kDa.Specifications
| TMEM27 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 0610008J07Rik; Cltrn; Collectrin; kidney-specific membrane protein; NX17; NX-17; Tmem27; Transmembrane protein 27; UNQ679/PRO1312 | |
| TMEM27 | |
| IgG2b κ | |
| Protein A/G | |
| Antibody |
| C5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9HBJ8 | |
| 57393 | |
| Cys17-Asp137 (Accession Q9HBJ8), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TMEM27 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title