missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM27 Monoclonal Antibody (C5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543559
This item is not returnable.
View return policy
Description
TMEM27 (Transmembrane protein 27) is also named as collectrin and Belongs to the TMEM27 family. It is a stimulator of beta cell replication and a membrane protein cleaved and shed by pancreatic beta cells that has been proposed as a beta cell mass biomarker. Western blot analysis of mouse collecting duct cell lines detected Tmem27 at an apparent molecular mass of 32 kDa. TMEM27 is an N-glycosylated protein and forms dimers, with MW of 64-75 kDa.
Specifications
| TMEM27 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9HBJ8 | |
| TMEM27 | |
| Cys17-Asp137 (Accession Q9HBJ8), with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| C5 | |
| Unconjugated | |
| TMEM27 | |
| 0610008J07Rik; Cltrn; Collectrin; kidney-specific membrane protein; NX17; NX-17; Tmem27; Transmembrane protein 27; UNQ679/PRO1312 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 57393 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction