missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM119 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TMEM119 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16825715
|
Proteintech
27585-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16815715
|
Proteintech
27585-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
27585-1-AP recognizes a 45-50 kDa band likely due to glycosylation.
TMEM119 immunohistochemistry might provide a useful tool for investigating the biology and pathology of human microglia. The predicted MW of TMEM119 is 29 kDa.Specifications
| TMEM119 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q4V9L6 | |
| 338773 | |
| TMEM119 Fusion Protein Ag26269 | |
| Primary | |
| -20°C | |
| TMEM119 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| OBIF, Osteoblast induction factor, TMEM119, transmembrane protein 119, UNQ731/PRO1415 | |
| TMEM119 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title