missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM119 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27585-1-AP-150UL
This item is not returnable.
View return policy
Description
27585-1-AP recognizes a 45-50 kDa band likely due to glycosylation.
TMEM119 immunohistochemistry might provide a useful tool for investigating the biology and pathology of human microglia. The predicted MW of TMEM119 is 29 kDa.Specifications
| TMEM119 | |
| Polyclonal | |
| Unconjugated | |
| TMEM119 | |
| OBIF, Osteoblast induction factor, TMEM119, transmembrane protein 119, UNQ731/PRO1415 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 338773 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4V9L6 | |
| TMEM119 | |
| TMEM119 Fusion Protein Ag26269 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction