missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM11 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93465-0.1ml
This item is not returnable.
View return policy
Description
TMEM11 Polyclonal antibody specifically detects TMEM11 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| TMEM11 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| C17orf35, chromosome 17 open reading frame 35, PM1putative receptor protein, PMI, Protein PM1, Protein PMI, transmembrane protein 11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human TMEM11 (NP_003867.1). MAAWGRRRLGPGSSGGSARERVSLSATDCYIVHEIYNGENAQDQFEYELEQALEAQYKYIVIEPTRIGDETARWITVGNC | |
| 0.1 mL | |
| Endocrinology, GPCR, Signal Transduction | |
| 8834 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction