missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMED9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TMED9 |
|---|---|
| Concentration | 0.26 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16811225
|
Proteintech
21620-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16801225
|
Proteintech
21620-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMED9 (also named as GMP25, GP25L2 or p25) is a member of the EMP24/GP25L family. TMED9 probably forms hetero-oligomeric complexes with TMED7 (p27), TMED2 (p24), and TMED10 (p23). Located in the endoplasmic reticulum and the Golgi, TMED9 appears to be involved in vesicular protein trafficking, mainly in the early secretory pathway.Specifications
| TMED9 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q5I0E7, Q99KF1, Q9BVK6 | |
| 361207, 54732, 67511 | |
| TMED9 Fusion Protein Ag13813 | |
| Primary | |
| -20°C | |
| TMED9 |
| 0.26 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Glycoprotein 25L2, GMP25, GP25L2, HSGP25L2G, TMED9 | |
| Tmed9 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title